Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Srgap1
Entrez Gene ID
117600
Gene Name
SLIT-ROBO Rho GTPase activating protein 1
Gene Aliases
4930572H05Rik, Arhgap13
Chromosome Location
Chr.10: 121780991 - 122047315 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081037.2 NP_001074506.1 8 - 9 1241 59
NM_001242411.1 NP_001229340.1 8 - 9 1241 59
GenBank mRNA AY057898.1 - 1 - 2 72 59
BC120892.1 - 8 - 9 1234 59
BC120893.1 - 8 - 9 1234 59
CD351674.1 - 4 - 5 588 59

View Full Product Details