Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Frmpd4
Entrez Gene ID
333605
Gene Name
FERM and PDZ domain containing 4
Gene Aliases
Gm196, PKAP1, Pdzd10, Pdzk10, Preso1, preso
Chromosome Location
Chr.X: 167471301 - 168577244 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033330.3 NP_001028502.1 10 - 11 1337 104
NM_001290427.1 NP_001277356.1 8 - 9 948 104
NM_001290428.1 NP_001277357.1 7 - 8 881 104
GenBank mRNA AK147370.1 - 10 - 11 1338 104
AK147440.1 - 7 - 8 882 104
BC139429.1 - 8 - 9 793 104

View Full Product Details