The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011165.3 | NP_035295.1 | 3 - 4 | 365 | 79 |
GenBank mRNA | AF011383.1 | - | 3 - 4 | 366 | 79 |
AF015562.1 | - | 3 - 4 | 365 | 79 | |
AK141285.1 | - | 3 - 4 | 377 | 79 | |
AK164009.1 | - | 3 - 4 | 377 | 79 | |
BC052218.1 | - | 3 - 4 | 366 | 79 |