Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nudcd1
Entrez Gene ID
67429
Gene Name
NudC domain containing 1
Gene Aliases
4921532K09Rik, AA407246, AW260430, AW556235, CML66-L, Cml66
Chromosome Location
Chr.15: 44375227 - 44428307 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001113554.1 NP_001107026.1 4 - 5 1159 78
NM_026149.4 NP_080425.3 4 - 5 1159 78
GenBank mRNA AF521132.1 - 4 - 5 1160 78
AK014996.2 - 4 - 5 1159 78
AK077673.1 - 4 - 5 741 78
AK083587.1 - 4 - 5 741 78
AK167855.1 - 4 - 5 691 78
AK168122.1 - 4 - 5 691 78
BC031583.1 - 4 - 5 725 78

View Full Product Details