Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nudcd1
Entrez Gene ID
67429
Gene Name
NudC domain containing 1
Gene Aliases
4921532K09Rik, AA407246, AW260430, AW556235, CML66-L, Cml66
Chromosome Location
Chr.15: 44375227 - 44428307 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001113554.1 NP_001107026.1 7 - 8 1702 132
NM_026149.4 NP_080425.3 7 - 8 1705 132
GenBank mRNA AF521132.1 - 7 - 8 1703 132
AK014996.2 - 7 - 8 1705 132
AK077673.1 - 7 - 8 1287 132
AK083587.1 - 7 - 8 1287 132
AK167855.1 - 7 - 8 1237 132
AK168122.1 - 7 - 8 1237 132
BC031583.1 - 7 - 8 1271 132

View Full Product Details