Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Klhl14
Entrez Gene ID
225266
Gene Name
kelch-like 14
Gene Aliases
6330403N15Rik, AI785250, printor
Chromosome Location
Chr.18: 21550377 - 21672932 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081403.1 NP_001074872.1 2 - 3 1067 89
GenBank mRNA AK018108.1 - 2 - 3 782 89
AK173163.1 - 2 - 3 781 89

View Full Product Details