Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nme5
Entrez Gene ID
75533
Gene Name
NME/NM23 family member 5
Gene Aliases
1700019D05Rik, Nm23-M5
Chromosome Location
Chr.18: 34562636 - 34579152 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_080637.3 NP_542368.2 2 - 3 238 137
GenBank mRNA AF343565.1 - 1 - 2 127 137
AK006111.1 - 1 - 2 222 137
AK131722.1 - 2 - 3 239 137
BC049625.1 - 2 - 3 232 137

View Full Product Details