Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Dsc3
Entrez Gene ID
13507
Gene Name
desmocollin 3
Gene Aliases
5430426I24Rik
Chromosome Location
Chr.18: 19960930 - 20002413 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001291809.1 NP_001278738.1 10 - 11 1685 70
NM_007882.3 NP_031908.3 10 - 11 1685 70
GenBank mRNA BC144845.1 - 10 - 11 1670 70
CD773111.1 - 2 - 3 338 70
Y11169.1 - 10 - 11 1685 70

View Full Product Details