Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Dsc3
Entrez Gene ID
13507
Gene Name
desmocollin 3
Gene Aliases
5430426I24Rik
Chromosome Location
Chr.18: 19960930 - 20002413 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001291809.1 NP_001278738.1 14 - 15 2416 98
NM_007882.3 NP_031908.3 14 - 15 2416 98
GenBank mRNA BC144845.1 - 14 - 15 2401 98

View Full Product Details