The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011397.4 | NP_035527.3 | 14 - 15 | 1928 | 190 |
GenBank mRNA | AK085499.1 | - | 14 - 15 | 1843 | 190 |
AK143959.1 | - | 14 - 15 | 1929 | 190 | |
AK165439.1 | - | 14 - 15 | 1864 | 190 | |
BC013528.1 | - | 14 - 15 | 1857 | 190 |