Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pfdn1
Entrez Gene ID
67199
Gene Name
prefoldin 1
Gene Aliases
2700086I23Rik, AA408327, AU044714
Chromosome Location
Chr.18: 36403679 - 36454495 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026027.3 NP_080303.1 2 - 3 203 81
GenBank mRNA AK002575.1 - 2 - 3 202 81
AK003330.1 - 2 - 3 221 81
AK010532.1 - 2 - 3 217 81
AK012575.1 - 2 - 3 204 81
AK028148.1 - 2 - 3 209 81
BC094627.1 - 2 - 3 204 81

View Full Product Details