Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Psd2
Entrez Gene ID
74002
Gene Name
pleckstrin and Sec7 domain containing 2
Gene Aliases
6330404E20Rik, AW125584, EFA6C
Chromosome Location
Chr.18: 35963881 - 36014715 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001289602.1 NP_001276531.1 14 - 15 2527 70
NM_028707.4 NP_082983.3 14 - 15 2533 70
NR_110349.1 14 - 15 2453 70
GenBank mRNA AK018116.1 - 9 - 10 989 70
AK034575.1 - 14 - 15 2303 70
AK045529.1 - 14 - 15 2264 70
AK053835.1 - 14 - 15 2181 70
AK149027.1 - 14 - 15 2263 70
BC056437.1 - 10 - 11 1032 70
BC062930.1 - 13 - 14 2114 70
BC065053.1 - 14 - 15 2301 70
BC065055.1 - 14 - 15 2284 70
BC066026.1 - 14 - 15 2533 70
BC066036.1 - 14 - 15 2290 70

View Full Product Details