The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001256382.1 | NP_001243311.1 | 23 - 24 | 4224 | 89 |
NM_001256383.1 | NP_001243312.1 | 22 - 23 | 4158 | 89 | |
NM_001256384.1 | NP_001243313.1 | 2 - 3 | 560 | 89 | |
NM_053271.2 | NP_444501.1 | 23 - 24 | 4098 | 89 | |
GenBank mRNA | AB021131.1 | - | 23 - 24 | 4052 | 89 |
AK032619.1 | - | 2 - 3 | 506 | 89 | |
AK129206.1 | - | 20 - 21 | 3322 | 89 | |
BC118979.2 | - | 22 - 23 | 4106 | 89 | |
HM015529.1 | - | 23 - 24 | 4147 | 89 |