Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tcerg1l
Entrez Gene ID
70571
Gene Name
transcription elongation regulator 1-like
Gene Aliases
5730476P14Rik
Chromosome Location
Chr.7: 138208972 - 138397730 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_183289.3 NP_899112.2 9 - 10 1487 83
GenBank mRNA BC048631.1 - 4 - 5 442 83

View Full Product Details