Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Zrsr2
- Entrez Gene ID
-
22184
- Gene Name
- zinc finger (CCCH type), RNA binding motif and serine/arginine rich 2
- Gene Aliases
-
35kDa, 5031411E02Rik, A230052C13Rik, C77286, U2af1-rs2, URP
- Chromosome Location
-
Chr.X: 163935443 - 163958820 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_009453.3 |
NP_033479.2 |
7 - 8 |
906 |
118 |
View Full Product Details