Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ace2
Entrez Gene ID
70008
Gene Name
angiotensin I converting enzyme (peptidyl-dipeptidase A) 2
Gene Aliases
2010305L05Rik
Chromosome Location
Chr.X: 164139342 - 164188418 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001130513.1 NP_001123985.1 4 - 5 770 69
NM_027286.4 NP_081562.2 3 - 4 622 69
GenBank mRNA AB053181.1 - 3 - 4 626 69
AB053182.1 - 3 - 4 626 69
AK141604.1 - 3 - 4 471 69
BC026801.1 - 3 - 4 615 69
EF408739.1 - 4 - 5 731 69
EF408740.1 - 4 - 5 731 69

View Full Product Details