The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001029872.3 | NP_001025043.3 | 24 - 25 | 2927 | 76 |
GenBank mRNA | AK132915.1 | - | - | 3071 | 76 |