Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nipal2
Entrez Gene ID
223473
Gene Name
NIPA-like domain containing 2
Gene Aliases
9330161F08Rik, Npal2
Chromosome Location
Chr.15: 34572799 - 34679437 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_145469.5 NP_663444.2 9 - 10 956 152
GenBank mRNA AK033975.1 - 9 - 10 957 152
AK038556.1 - 9 - 10 1098 152
AK079085.1 - 9 - 10 1388 152
BC016107.1 - 9 - 10 979 152
BC030399.1 - 9 - 10 1473 152
BC058207.1 - 9 - 10 1122 152

View Full Product Details