Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cntnap2
Entrez Gene ID
66797
Gene Name
contactin associated protein-like 2
Gene Aliases
5430425M22Rik, Caspr2, mKIAA0868
Chromosome Location
Chr.6: 45060061 - 47301388 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025771.3 NP_080047.1 1 - 2 153
GenBank mRNA AK013139.1 - 1 - 2 153
AK017341.1 - 1 - 2 153
BC029826.1 - 1 - 2 104 153
BY297045.1 - 1 - 2 153

View Full Product Details