The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_029202.2 | NP_083478.2 | 2 - 3 | 528 | 144 |
GenBank mRNA | AK015948.1 | - | 2 - 3 | 472 | 144 |
BC049731.1 | - | 2 - 3 | 528 | 144 |