The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001191012.1 | NP_001177941.1 | 5 - 6 | 637 | 76 |
GenBank mRNA | GU269270.1 | - | 5 - 6 | 623 | 76 |