The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_028873.4 | NP_083149.3 | 6 - 7 | 1987 | 119 |
GenBank mRNA | AK017830.1 | - | 6 - 7 | 2001 | 119 |
AK032681.1 | - | 6 - 7 | 1969 | 119 | |
AK045345.1 | - | 6 - 7 | 2044 | 119 | |
AK045445.1 | - | 6 - 7 | 2081 | 119 | |
AK082579.1 | - | 6 - 7 | 1988 | 119 | |
AK083878.1 | - | 6 - 7 | 1999 | 118 | |
AK159388.1 | - | 6 - 7 | 2000 | 119 | |
AK168935.1 | - | 6 - 7 | 2079 | 119 | |
BC011146.1 | - | 6 - 7 | 2052 | 119 |