Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Slc17a4
Entrez Gene ID
319848
Gene Name
solute carrier family 17 (sodium phosphate), member 4
Gene Aliases
9130214H05Rik
Chromosome Location
Chr.13: 23897879 - 23915009 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_177016.3 NP_795990.2 7 - 8 928 97
GenBank mRNA AK033669.1 - 7 - 8 931 97
AK162320.1 - 6 - 7 1371 97
BC147415.1 - 7 - 8 889 97
BC147416.1 - 7 - 8 889 97

View Full Product Details