Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tas2r115
Entrez Gene ID
353325
Gene Name
taste receptor, type 2, member 115
Gene Aliases
T2R15, Tas2r15, mGR15, mt2r49
Chromosome Location
Chr.6: 132737054 - 132737986 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_207020.1 NP_996903.1 1 - 1 834 105
GenBank mRNA BC138393.1 - 1 - 1 883 105
BC138398.1 - 1 - 1 883 105

View Full Product Details