Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Arid4b
Entrez Gene ID
94246
Gene Name
AT rich interactive domain 4B (RBP1-like)
Gene Aliases
BCAA, BRCAA1, RBBP1L1, Rbp1l1, SAP180
Chromosome Location
Chr.13: 14063768 - 14199603 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_194262.2 NP_919238.1 14 - 15 1464 104
NM_198122.2 NP_937755.1 14 - 15 1464 104
GenBank mRNA AK031062.1 - 14 - 15 1447 104
AK041414.1 - 14 - 15 1461 104
AK080355.1 - 4 - 5 324 104
AK136105.1 - 14 - 15 1348 104
AK161669.1 - 14 - 15 1450 104
BC137783.1 - 13 - 14 1226 104

View Full Product Details