Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tfec
Entrez Gene ID
21426
Gene Name
transcription factor EC
Gene Aliases
BB107417, Tcfec, bHLHe34
Chromosome Location
Chr.6: 16830837 - 16898462 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031198.3 NP_112475.1 3 - 4 546 108
GenBank mRNA AF077742.1 - 3 - 4 546 108
AK137141.1 - 3 - 4 511 108
AK156327.1 - 3 - 4 459 108
AK159579.1 - 3 - 4 479 108
BC098494.1 - 3 - 4 431 108

View Full Product Details