Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tfec
Entrez Gene ID
21426
Gene Name
transcription factor EC
Gene Aliases
BB107417, Tcfec, bHLHe34
Chromosome Location
Chr.6: 16830837 - 16898462 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031198.3 NP_112475.1 4 - 5 603 116
GenBank mRNA AF077742.1 - 4 - 5 603 116
AK137141.1 - 4 - 5 568 116
AK156327.1 - 4 - 5 516 116
AK159579.1 - 4 - 5 536 116
BC098494.1 - 4 - 5 488 116

View Full Product Details