The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_027994.1 | NP_082270.1 | 8 - 9 | 1636 | 60 |
GenBank mRNA | AK129225.1 | - | 8 - 9 | 1597 | 60 |
AK144869.1 | - | 8 - 9 | 1628 | 60 | |
AK158080.1 | - | 8 - 9 | 1632 | 60 | |
BC057457.1 | - | 1 - 2 | 279 | 60 |