Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Trim27
Entrez Gene ID
19720
Gene Name
tripartite motif-containing 27
Gene Aliases
AW538890, Gm19403, Rfp
Chromosome Location
Chr.13: 21179440 - 21194723 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009054.3 NP_033080.2 6 - 7 1479 83
GenBank mRNA AK031744.1 - 6 - 7 1197 83
AK088180.1 - 6 - 7 963 83
BC003219.1 - 6 - 7 984 83
BC069924.1 - 6 - 7 1190 83
L46855.1 - - 1090 83
X75343.1 - 6 - 7 894 83

View Full Product Details