Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Mrpl48
- Entrez Gene ID
-
52443
- Gene Name
- mitochondrial ribosomal protein L48
- Gene Aliases
-
1810030E20Rik, 2610028L11Rik, CGI-118, D4Ertd786e
- Chromosome Location
-
Chr.7: 100548706 - 100607996 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BE995503.1 |
- |
2 - 3 |
163 |
114 |
View Full Product Details