Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Chrdl2
Entrez Gene ID
69121
Gene Name
chordin-like 2
Gene Aliases
1810022C01Rik, Chl2
Chromosome Location
Chr.7: 100006404 - 100034728 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001291320.1 NP_001278249.1 1 - 2 202 117
GenBank mRNA AK007577.1 - 1 - 2 206 117

View Full Product Details