Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sema5a
Entrez Gene ID
20356
Gene Name
sema domain, seven thrombospondin repeats (type 1 and type 1-like), transmembrane domain (TM) and short cytoplasmic domain, (semaphorin) 5A
Gene Aliases
5930434A13, 9130201M22Rik, AI464145, Semaf, semF
Chromosome Location
Chr.15: 32244742 - 32696341 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009154.2 NP_033180.2 9 - 10 1587 82
GenBank mRNA AK031231.1 - 9 - 10 1580 82
AK043386.1 - 9 - 10 1299 82
AK046502.1 - 9 - 10 1363 82
AK141612.1 - 9 - 10 1531 82
AK143026.1 - 9 - 10 1598 82
AK157706.1 - 9 - 10 1570 82
BC065137.1 - 8 - 9 1056 82
X97817.1 - 7 - 8 956 82

View Full Product Details