Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ppp1r9a
Entrez Gene ID
243725
Gene Name
protein phosphatase 1, regulatory (inhibitor) subunit 9A
Gene Aliases
2810430P21Rik, 4930518N04Rik, 5330407E15, A230094E16Rik, BB181831, NRB, mKIAA1222, neurabin-I
Chromosome Location
Chr.6: 4902747 - 5165661 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_181595.3 NP_853626.1 13 - 14 3232 72
GenBank mRNA AB091828.1 - 13 - 14 3232 72
AK122472.1 - 5 - 6 783 72
AK138313.1 - 13 - 14 3161 72
AY308065.1 - 12 - 13 2907 72

View Full Product Details