Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Scnn1g
Entrez Gene ID
20278
Gene Name
sodium channel, nonvoltage-gated 1 gamma
Gene Aliases
[g]ENaC
Chromosome Location
Chr.7: 121734420 - 121768477 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011326.2 NP_035456.1 7 - 8 1261 96
GenBank mRNA AF112187.1 - 7 - 8 1261 96
AK143995.1 - 6 - 7 954 96
BC021338.1 - 7 - 8 1300 96

View Full Product Details