Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tnnt3
Entrez Gene ID
21957
Gene Name
troponin T3, skeletal, fast
Gene Aliases
fTnT
Chromosome Location
Chr.7: 142498777 - 142516213 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163664.1 NP_001157136.1 3 - 4 109 63
NM_001163665.1 NP_001157137.1 3 - 4 109 63
NM_001163666.1 NP_001157138.1 3 - 4 109 63
NM_001163667.1 NP_001157139.1 3 - 4 109 63
NM_001163668.1 NP_001157140.1 3 - 4 109 63
NM_001163669.1 NP_001157141.1 3 - 4 109 63
GenBank mRNA AA028290.1 - 3 - 4 89 63
AA771596.1 - 3 - 4 87 63
AI173874.1 - 3 - 4 101 63
AI574395.1 - 3 - 4 103 63
AI746973.1 - 3 - 4 104 63
BC003747.1 - 3 - 4 105 63
BY766323.1 - 3 - 4 99 63
L49466.1 - 3 - 4 109 63
L49467.1 - 3 - 4 109 63
L49468.1 - 3 - 4 109 63
L49469.1 - 3 - 4 109 63
L49470.1 - 3 - 4 109 63
L49472.1 - 3 - 4 109 63

View Full Product Details