Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tnnt3
Entrez Gene ID
21957
Gene Name
troponin T3, skeletal, fast
Gene Aliases
fTnT
Chromosome Location
Chr.7: 142498777 - 142516213 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163669.1 NP_001157141.1 5 - 6 154 68
GenBank mRNA AA028290.1 - 5 - 6 134 68
L49470.1 - 5 - 6 154 68

View Full Product Details