Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tnnt3
Entrez Gene ID
21957
Gene Name
troponin T3, skeletal, fast
Gene Aliases
fTnT
Chromosome Location
Chr.7: 142498777 - 142516213 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163667.1 NP_001157139.1 6 - 7 165 114
NM_001163668.1 NP_001157140.1 6 - 7 165 75
GenBank mRNA AA771596.1 - 6 - 7 143 75
BY766323.1 - 6 - 7 155 114
L49466.1 - 6 - 7 165 114
L49471.1 - 5 - 6 147 114
L49472.1 - 6 - 7 165 75

View Full Product Details