Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc25a32
Entrez Gene ID
69906
Gene Name
solute carrier family 25, member 32
Gene Aliases
2610043O12Rik, Mftc
Chromosome Location
Chr.15: 39094191 - 39112768 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172402.3 NP_765990.2 6 - 7 1024 116
GenBank mRNA AK031187.1 - 6 - 7 829 116
AK145178.1 - 6 - 7 984 116
AK167501.1 - 6 - 7 834 116
AK170643.1 - 6 - 7 993 116
AK172029.1 - 6 - 7 992 116
BC031874.1 - 6 - 7 958 116

View Full Product Details