The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001102458.1 | NP_001095928.1 | 11 - 12 | 1764 | 80 |
NM_001301688.1 | NP_001288617.1 | 10 - 11 | 1604 | 80 | |
NM_018745.5 | NP_061215.1 | 10 - 11 | 1604 | 80 | |
NR_125913.1 | 10 - 11 | 1753 | 80 | ||
GenBank mRNA | AF032128.1 | - | 8 - 9 | 1030 | 80 |
AK049680.1 | - | 10 - 11 | 1605 | 80 | |
AK088112.1 | - | 10 - 11 | 1602 | 80 | |
AK088671.1 | - | 11 - 12 | 1761 | 80 | |
AK133914.1 | - | 10 - 11 | 1600 | 80 | |
BC019412.1 | - | 10 - 11 | 1592 | 80 | |
BC021152.1 | - | 10 - 11 | 1557 | 80 | |
BC043722.1 | - | 8 - 9 | 1110 | 80 | |
BC046814.1 | - | 10 - 11 | 1656 | 80 | |
CF738062.1 | - | 6 - 7 | 595 | 80 | |
CF749918.1 | - | 1 - 2 | 57 | 80 |