The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_028758.2 | NP_083034.1 | 1 - 2 | 208 | 88 |
GenBank mRNA | AK004632.1 | - | 1 - 2 | 180 | 88 |
AK129280.1 | - | 1 - 2 | 167 | 88 | |
AK143691.1 | - | 1 - 2 | 196 | 88 | |
AK148205.1 | - | - | 191 | 88 | |
AK149798.1 | - | 1 - 2 | 198 | 88 | |
AK154904.1 | - | 1 - 2 | 183 | 88 | |
AK159953.1 | - | 1 - 2 | 198 | 88 | |
BC062933.1 | - | 1 - 2 | 177 | 88 | |
BY291970.1 | - | 1 - 2 | 208 | 88 |