Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cog7
Entrez Gene ID
233824
Gene Name
component of oligomeric golgi complex 7
Gene Aliases
5630400E24Rik, Gm167
Chromosome Location
Chr.7: 121922836 - 121981706 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033318.3 NP_001028490.2 11 - 12 1578 92
GenBank mRNA AK030709.1 - 11 - 12 1545 92
AK145163.1 - 11 - 12 1576 92
AK154693.1 - 11 - 12 1580 92
AK163143.1 - - 1598 92
AK172200.1 - 11 - 12 1580 92

View Full Product Details