Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cog7
Entrez Gene ID
233824
Gene Name
component of oligomeric golgi complex 7
Gene Aliases
5630400E24Rik, Gm167
Chromosome Location
Chr.7: 121922836 - 121981706 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033318.3 NP_001028490.2 7 - 8 1119 81
GenBank mRNA AK030709.1 - 7 - 8 1086 81
AK145163.1 - 7 - 8 1117 81
AK154693.1 - 7 - 8 1121 81
AK163143.1 - - 1139 81
AK172200.1 - 7 - 8 1121 81

View Full Product Details