Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Acsm5
Entrez Gene ID
272428
Gene Name
acyl-CoA synthetase medium-chain family member 5
Gene Aliases
C730019D22, C730027J19Rik
Chromosome Location
Chr.7: 119526131 - 119543360 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_178758.3 NP_848873.1 6 - 7 1115 66
GenBank mRNA AK050132.1 - 7 - 8 1212 66
AK050219.1 - 6 - 7 1140 66
AK050288.1 - 6 - 7 1135 66
AK050404.1 - 6 - 7 1175 66
AK050458.1 - 6 - 7 1077 66
AK149591.1 - 6 - 7 1135 66
BC095985.1 - 6 - 7 1115 66

View Full Product Details