Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Acsm5
Entrez Gene ID
272428
Gene Name
acyl-CoA synthetase medium-chain family member 5
Gene Aliases
C730019D22, C730027J19Rik
Chromosome Location
Chr.7: 119526131 - 119543360 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_178758.3 NP_848873.1 8 - 9 1315 90
GenBank mRNA AK050132.1 - 9 - 10 1412 90
AK050219.1 - 8 - 9 1340 90
AK050288.1 - 8 - 9 1335 90
AK050404.1 - 8 - 9 1375 90
AK050458.1 - 8 - 9 1277 90
AK149591.1 - 8 - 9 1335 90
BC095985.1 - 8 - 9 1315 90

View Full Product Details