The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_027164.1 | NP_081440.1 | 6 - 7 | 1268 | 125 |
GenBank mRNA | BC050816.1 | - | 6 - 7 | 1268 | 125 |