The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001177961.1 | NP_001171432.1 | 1 - 2 | 201 | 59 |
NM_001177962.1 | NP_001171433.1 | 1 - 2 | 201 | 59 | |
NM_023122.3 | NP_075611.1 | 1 - 2 | 201 | 59 | |
GenBank mRNA | AF254871.1 | - | 1 - 2 | 172 | 59 |
AF254876.1 | - | 1 - 2 | 172 | 59 | |
AF254877.1 | - | 1 - 2 | 172 | 59 | |
AK016567.1 | - | 1 - 2 | 126 | 59 | |
AK132778.1 | - | 1 - 2 | 188 | 59 | |
BC025165.1 | - | 1 - 2 | 162 | 59 | |
CJ155269.1 | - | 1 - 2 | 201 | 59 |