Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lhfpl3
Entrez Gene ID
269629
Gene Name
lipoma HMGIC fusion partner-like 3
Gene Aliases
A930031L14Rik
Chromosome Location
Chr.5: 22745636 - 23275597 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK076730.1 - 1 - 2 104 89

View Full Product Details