Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Celf4
Entrez Gene ID
108013
Gene Name
CUGBP, Elav-like family member 4
Gene Aliases
A230070D14Rik, BRUNOL-4, Brul4, Brunol4, C130060B05Rik
Chromosome Location
Chr.18: 25477620 - 25754162 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001146292.1 NP_001139764.1 1 - 2 647 72
NM_001146293.1 NP_001139765.1 1 - 2 647 72
NM_001146294.1 NP_001139766.1 1 - 2 647 72
NM_001146295.1 NP_001139767.1 1 - 2 647 72
NM_001174074.1 NP_001167545.1 1 - 2 647 72
NM_133195.3 NP_573458.2 1 - 2 647 72
GenBank mRNA AF515450.1 - 1 - 2 415 72
AK044072.1 - 1 - 2 337 72
AK048427.1 - 1 - 2 824 72
AK051458.1 - 1 - 2 368 72
AK161918.1 - 1 - 2 618 72
BC048405.1 - 1 - 2 647 72
BC052744.1 - 1 - 2 647 72

View Full Product Details