Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fhod3
Entrez Gene ID
225288
Gene Name
formin homology 2 domain containing 3
Gene Aliases
A930009H06Rik, FHOS2, mKIAA1695
Chromosome Location
Chr.18: 24708456 - 25133507 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001289654.1 NP_001276583.1 22 - 23 4854 65
NM_001289655.1 NP_001276584.1 23 - 24 5211 65
NM_175276.4 NP_780485.2 24 - 25 5307 65
GenBank mRNA AB078608.1 - 24 - 25 4485 65
AB078609.1 - 22 - 23 4032 65
AB100088.1 - 23 - 24 4389 65
AK220413.1 - 20 - 21 3471 65
BC056371.1 - 14 - 15 2937 65
BC060607.1 - 9 - 10 2263 65
BC138046.1 - 22 - 23 3954 65

View Full Product Details