Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Eny2
- Entrez Gene ID
-
223527
- Gene Name
- enhancer of yellow 2 homolog (Drosophila)
- Gene Aliases
-
1810057B09Rik, 6720481I12, DC6, Ey2
- Chromosome Location
-
Chr.15: 44428081 - 44438256 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK016562.1 |
- |
1 - 2 |
1486 |
105 |
View Full Product Details